Assay Details
Gene Symbol: GPR160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014373.2 | NP_055188.1 | 1 - 2 | 269 | 123 | |
XM_005247346.3 | XP_005247403.1 | 1 - 2 | 78 | 123 | ||
XM_017006161.1 | XP_016861650.1 | 1 - 2 | 78 | 123 | ||
XM_017006162.1 | XP_016861651.1 | 1 - 2 | 218 | 123 | ||
XM_017006163.1 | XP_016861652.1 | 1 - 2 | 893 | 123 | ||
GenBank mRNA | AK311293.1 | - | 1 - 2 | 78 | 123 | |
BC000181.2 | - | 1 - 2 | 204 | 3048193 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000910.hg
Additional Information:
This assay is in the 5'UTR region of the BC000181.2,NM_014373.2,XM_005247346.3,XM_017006161.1,XM_017006162.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03015914.hg.1 | ENST00000355897.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015914.hg.1 | ENST00000482813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015914.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015914.hg.1 | TC03000910.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015907.hg.1 | ENST00000355897.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015907.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015907.hg.1 | TC03000910.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015908.hg.1 | ENST00000355897.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015908.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015908.hg.1 | TC03000910.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015909.hg.1 | ENST00000355897.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015909.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015909.hg.1 | TC03000910.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0300167949.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167949.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167949.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167949.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167949.hg.1 | TC0300009459.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167950.hg.1 | TC0300009459.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167955.hg.1 | TC0300009459.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167948.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167948.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167948.hg.1 | ENST00000355897.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167948.hg.1 | NM_014373.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167948.hg.1 | TC0300009459.hg.1 | Clariom D TC |
