This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00977895_g1
See other CDK9 GE Assays ›
Gene Symbol:
CDK9
Gene Name:
cyclin dependent kinase 9
Gene Aliases:
C-2k, CDC2L4, CTK1, PITALRE, TAK
Chromosome Location:
Chr.9: 127786026 - 127790782 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00977895_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDK9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261.3 NP_001252.1 5 - 6 731 103
XM_017014184.1 XP_016869673.1 5 - 6 791 103
GenBank mRNA AB451289.1 - 5 - 6 608 103
AB451421.1 - 5 - 6 608 103
AK313828.1 - 5 - 6 660 103
BC001968.1 - 5 - 6 668 3460767 103
BT019903.1 - 5 - 6 608 103
L25676.1 - 5 - 6 672 103
X80230.1 - 5 - 6 706 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900012175.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000671.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDK9

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
regulation of DNA repair
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
protein phosphorylation
regulation of mitotic cell cycle
cell proliferation
positive regulation of cardiac muscle hypertrophy
regulation of histone modification
replication fork processing
positive regulation of histone phosphorylation
response to drug
snRNA transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of viral transcription
regulation of muscle cell differentiation
negative regulation of cell cycle arrest
cellular response to cytokine stimulus
negative regulation of mRNA polyadenylation
positive regulation of mRNA 3'-UTR binding
positive regulation of histone H2B ubiquitination
RNA polymerase II core promoter sequence-specific DNA binding
transcription coactivator binding
DNA binding
chromatin binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
kinase activity
7SK snRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09008020.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09008020.hg.1 ENST00000373265.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09008020.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09008020.hg.1 TC09000671.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09008021.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09008021.hg.1 ENST00000373265.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09008021.hg.1 NM_001261.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09008021.hg.1 TC09000671.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000480353.5 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay TC0900012175.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 ENST00000491521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay PSR0900184734.hg.1 TC0900012175.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 ENST00000373264.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 ENST00000498339.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 NM_001261.3 Refseq
Affymetrix Human Clariom D Assay PSR0900184735.hg.1 TC0900012175.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0