This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00977759_m1
See other EFTUD2 GE Assays ›
Gene Symbol:
EFTUD2
Gene Name:
elongation factor Tu GTP binding domain containing 2
Gene Aliases:
MFDGA, MFDM, SNRNP116, Snrp116, Snu114, U5-116KD
Chromosome Location:
Chr.17: 44850287 - 44899625 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00977759_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EFTUD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142605.1 NP_001136077.1 24 - 25 2714 76
NM_001258353.1 NP_001245282.1 25 - 26 2960 76
NM_001258354.1 NP_001245283.1 25 - 26 2793 76
NM_004247.3 NP_004238.3 25 - 26 2823 76
GenBank mRNA AJ505017.1 - 21 - 22 2197 76
AK126193.1 - 14 - 15 2923 76
AK126464.1 - 26 - 27 2629 76
AK126652.1 - 8 - 9 2854 76
AK128148.1 - 25 - 26 2961 76
AK225335.1 - 25 - 26 2583 76
AK293108.1 - 25 - 26 2586 76
AK296367.1 - 24 - 25 2517 76
AK297392.1 - 25 - 26 2599 76
AK302828.1 - 25 - 26 2463 76
AK316098.1 - 24 - 25 2534 76
BC002360.2 - 25 - 26 2593 2960986 76
CR456774.1 - 24 - 25 2562 76
D21163.1 - 25 - 26 2622 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700010849.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001586.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EFTUD2

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
mRNA processing
translation
RNA splicing
cellular response to drug
response to cocaine
GTPase activity
protein binding
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17021348.hg.1 ENST00000262414.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17021348.hg.1 ENST00000402521.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17021348.hg.1 ENST00000426333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17021348.hg.1 NM_001142605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17021348.hg.1 NM_004247.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17021348.hg.1 TC17001586.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000402521.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000426333.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000586276.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000589769.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000590367.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000590977.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000591382.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000592576.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_001142605.1 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_001258353.1 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_001258354.1 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_004247.3 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000402521.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000426333.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000586276.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000589769.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000590367.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000590977.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000591382.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 ENST00000592576.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_001142605.1 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_001258353.1 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_001258354.1 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 NM_004247.3 Refseq
Affymetrix Human Clariom D Assay PSR1700186870.hg.1 TC1700010849.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0