This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00975912_m1
See other MED12L GE Assays ›
Gene Symbol:
MED12L
Gene Name:
mediator complex subunit 12 like
Gene Aliases:
NOPAR, TNRC11L, TRALP, TRALPUSH
Chromosome Location:
Chr.3: 151085665 - 151436677 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00975912_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MED12L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053002.5 NP_443728.3 28 - 29 4338 95
XM_006713487.3 XP_006713550.1 30 - 31 4715 95
XM_011512390.2 XP_011510692.1 29 - 30 4610 95
XM_011512394.2 XP_011510696.1 30 - 31 4715 95
XM_017005676.1 XP_016861165.1 30 - 31 4715 95
XM_017005677.1 XP_016861166.1 30 - 31 4712 95
XM_017005678.1 XP_016861167.1 30 - 31 4715 95
XM_017005679.1 XP_016861168.1 29 - 30 4445 95
XM_017005680.1 XP_016861169.1 28 - 29 4433 95
XM_017005681.1 XP_016861170.1 15 - 16 2101 95
GenBank mRNA AB046855.1 - 15 - 16 2081 95
AF090917.1 - 5 - 6 703 95
AF399708.1 - 28 - 29 4247 95
AK022714.1 - 12 - 13 1787 95
AK128165.1 - 12 - 13 1927 95
BC144559.1 - 26 - 27 3822 9053087 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300009179.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000822.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MED12L

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription coactivator activity
beta-catenin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000273432.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000309237.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000363124.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000422248.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000468305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000468305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000468305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000469768.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000469768.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000469768.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000474524.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000480026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000480026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000488092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000488092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000491549.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000491549.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000491549.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000491549.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000491549.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 ENST00000491549.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009179.hg.1 NM_053002.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0