This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00974249_g1
See other MMP15 GE Assays ›
Gene Symbol:
MMP15
Gene Name:
matrix metallopeptidase 15
Gene Aliases:
MMP-15, MT2-MMP, MT2MMP, MTMMP2, SMCP-2
Chromosome Location:
Chr.16: 58025566 - 58046901 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00974249_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MMP15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002428.3 NP_002419.1 6 - 7 1951 68
GenBank mRNA BC006412.1 - 6 - 7 1071 68
BC036495.1 - 6 - 7 1957 5263145 68
BC055428.1 - 6 - 7 1952 4811572 68
BT006715.1 - 4 - 5 851 68
D85510.1 - 3 - 4 681 68
D86331.1 - 5 - 6 889 68
Z48482.1 - 6 - 7 1214 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008021.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000500.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MMP15

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
proteolysis
extracellular matrix disassembly
collagen catabolic process
response to estradiol
endodermal cell differentiation
positive regulation of catalytic activity
metalloendopeptidase activity
serine-type endopeptidase activity
calcium ion binding
protein binding
enzyme activator activity
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16007017.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16007017.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007017.hg.1 TC16000500.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000570065.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 ENST00000570065.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000570065.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 ENST00000570065.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay TC1600008021.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay PSR1600159213.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600159213.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay PSR1600159213.hg.1 ENST00000219271.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600159213.hg.1 NM_002428.3 Refseq
Affymetrix Human Clariom D Assay PSR1600159213.hg.1 TC1600008021.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0