This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00974100_s1
See other KCNMA1 GE Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00974100_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNMA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017016221.1 XP_016871710.1 2 1037 94
GenBank mRNA AK098326.1 - 2 2501 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB351610.1 - 1 Chr. 10 - 77634111 - 77634613 [+] on Build GRCh38 94
GenBankmRNA H29316.1 - 1 Chr. 10 - 77634110 - 77634466 [+] on Build GRCh38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream KCNMA1 antisense RNA 3 KCNMA1-AS3 -257.5kb Sense Chr. 10 - 77350872 - 77376613 on NCBI Build GRCh38
Downstream discs large MAGUK scaffold protein 5 DLG5 LP-DLG,
P-DLG5,
PDLG
156.3kb Anti-sense Chr. 10 - 77790787 - 77926590 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0