This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00973138_m1
See other MAGEC3 GE Assays ›
Gene Symbol:
MAGEC3
Gene Name:
MAGE family member C3
Gene Aliases:
CT7.2, HCA2, MAGE-C3, MAGEC4
Chromosome Location:
Chr.X: 141838316 - 141897832 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00973138_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAGEC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138702.1 NP_619647.1 6 - 7 1118 56
GenBank mRNA AF490508.1 - 6 - 7 1118 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008611.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000691.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MAGEC3

Panther Classification:

Molecular Function -

scaffold/adaptor protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 ENST00000536088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 ENST00000544766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008915.hg.1 TC0X000691.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 ENST00000483584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 ENST00000536088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 ENST00000544766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 NM_138702.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 NM_177456.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008914.hg.1 TC0X000691.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00158477.hg.1 TC0X00008611.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00158476.hg.1 TC0X00008611.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000298296.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000409007.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000443323.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000483584.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 ENST00000544766.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_138702.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq
Affymetrix Human Clariom D Assay TC0X00008611.hg.1 NM_177456.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header