Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000306270.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000369751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000503400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000503631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000505222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000510291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000610980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
NM_001300906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
NM_015525.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000306270.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000369751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000503400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000503631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000505222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000510291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
ENST00000610980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
NM_001300906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
NM_015525.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185469.hg.1 |
TC0600012413.hg.1 |
Clariom D TC |