This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00970644_m1
See other NEK5 GE Assays ›
Gene Symbol:
NEK5
Gene Name:
NIMA related kinase 5
Gene Aliases:
-
Chromosome Location:
Chr.13: 52064762 - 52129092 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00970644_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEK5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199289.2 NP_954983.1 9 - 10 766 135
XM_006719808.3 XP_006719871.2 9 - 10 847 135
XM_011535066.2 XP_011533368.1 9 - 10 847 135
XM_011535068.2 XP_011533370.1 9 - 10 847 135
XM_017020555.1 XP_016876044.1 9 - 10 849 135
XM_017020556.1 XP_016876045.1 9 - 10 853 135
XM_017020557.1 XP_016876046.1 9 - 10 857 135
GenBank mRNA BC063885.1 - 9 - 10 752 30383658 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300010040.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000685.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NEK5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
positive regulation of striated muscle cell differentiation
positive regulation of cysteine-type endopeptidase activity
protein serine/threonine kinase activity
ATP binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000355568.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 ENST00000617045.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300010040.hg.1 NM_199289.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header