Assay Details
Gene Symbol: DENND6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001794.3 | NP_001001794.3 | 9 - 10 | 833 | 84 | |
XM_005261914.3 | XP_005261971.1 | 10 - 11 | 1008 | 84 | ||
XM_011530692.2 | XP_011528994.1 | 10 - 11 | 1008 | 84 | ||
XM_011530693.2 | XP_011528995.1 | 9 - 10 | 787 | 84 | ||
XM_017028801.1 | XP_016884290.1 | 8 - 9 | 844 | 84 | ||
GenBank mRNA | AK054743.1 | - | 7 - 8 | 952 | 84 | |
BC031069.1 | - | 9 - 10 | 833 | 5296726 | 84 | |
BX363595.2 | - | 9 - 10 | 823 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000912.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22015489.hg.1 | ENST00000413817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015489.hg.1 | ENST00000471942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015489.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22015489.hg.1 | TC22000912.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000460087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009153.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000413817.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000433760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000471942.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | ENST00000495607.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | NM_001001794.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200166377.hg.1 | TC2200009153.hg.1 | Clariom D TC |
