This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00970127_g1
See other ALG12 GE Assays ›
Gene Symbol:
ALG12
Gene Name:
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases:
CDG1G, ECM39, PP14673, hALG12
Chromosome Location:
Chr.22: 49860163 - 49918469 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00970127_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALG12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024105.3 NP_077010.1 2 - 3 438 81
XM_011530371.2 XP_011528673.1 2 - 3 449 81
XM_017028936.1 XP_016884425.1 2 - 3 449 81
XM_017028937.1 XP_016884426.1 2 - 3 449 81
GenBank mRNA AF318343.1 - 2 - 3 390 81
AJ290427.1 - 2 - 3 311 81
AJ303120.1 - 2 - 3 311 81
BC001729.1 - 2 - 3 285 3353085 81
BC098562.1 - 2 - 3 331 30560020 81
CB858085.1 - 2 - 3 439 81
CR456369.1 - 1 - 2 227 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200009128.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000904.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALG12

Panther Classification:

Molecular Function -

glycosyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein folding
protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
mannosylation
alpha-1,6-mannosyltransferase activity
mannosyltransferase activity
dol-P-Man:Man(7)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22015112.hg.1 ENST00000330817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015112.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015112.hg.1 TC22000904.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22015114.hg.1 ENST00000330817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015114.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015114.hg.1 TC22000904.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22015113.hg.1 ENST00000330817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015113.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015113.hg.1 TC22000904.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22015116.hg.1 ENST00000330817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015116.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015116.hg.1 TC22000904.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015115.hg.1 ENST00000330817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015115.hg.1 NM_024105.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015115.hg.1 TC22000904.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2200165970.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165970.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165970.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165970.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165970.hg.1 TC2200009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200165972.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165972.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165972.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165972.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165972.hg.1 TC2200009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200165973.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165973.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165973.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165973.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165973.hg.1 TC2200009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000486602.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 ENST00000492791.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay TC2200009128.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165971.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165971.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165971.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165971.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165971.hg.1 TC2200009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200165974.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165974.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165974.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165974.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165974.hg.1 TC2200009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200165969.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165969.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165969.hg.1 ENST00000330817.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200165969.hg.1 NM_024105.3 Refseq
Affymetrix Human Clariom D Assay PSR2200165969.hg.1 TC2200009128.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 112798 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0