Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017122.hg.1 |
ENST00000375262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017122.hg.1 |
ENST00000375263.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017122.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017122.hg.1 |
TC09001372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017123.hg.1 |
ENST00000375263.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017123.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017123.hg.1 |
TC09001372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017121.hg.1 |
ENST00000375262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017121.hg.1 |
ENST00000375263.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017121.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017121.hg.1 |
TC09001372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000467499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000467499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000467499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000484816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000484816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174207.hg.1 |
TC0900010907.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000463517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000464104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000467499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000467499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000467499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000484816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000484816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
ENST00000494814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010907.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174208.hg.1 |
TC0900010907.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
ENST00000375262.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
ENST00000375263.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
NM_000197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174206.hg.1 |
TC0900010907.hg.1 |
Clariom D TC |