This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00968489_m1
See other HAT1 GE Assays ›
Gene Symbol:
HAT1
Gene Name:
histone acetyltransferase 1
Gene Aliases:
KAT1
Chromosome Location:
Chr.2: 171922425 - 171983686 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00968489_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003642.3 NP_003633.1 3 - 4 266 146
XM_006712808.3 XP_006712871.1 4 - 5 462 146
XM_011512027.2 XP_011510329.1 3 - 4 1018 146
XM_017005136.1 XP_016860625.1 4 - 5 1239 146
XM_017005137.1 XP_016860626.1 4 - 5 1171 146
GenBank mRNA AF030424.1 - 3 - 4 226 146
AK313699.1 - 3 - 4 257 147
DB050336.1 - 4 - 5 467 146

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009964.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001024.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HAT1

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

DNA packaging
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
chromatin silencing at telomere
internal protein amino acid acetylation
response to nutrient
chromatin modification
histone H4 acetylation
histone acetyltransferase activity
protein binding
H4 histone acetyltransferase activity
histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0