This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00965871_m1
See other AGPAT1 GE Assays ›
Gene Symbol:
AGPAT1
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases:
1-AGPAT1, G15, LPAAT-alpha, LPAATA
Chromosome Location:
Chr.6: 32168206 - 32178111 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00965871_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGPAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011514234.1 XP_011512536.1 1 - 2 294 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011514234.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
positive regulation of cellular metabolic process
1-acylglycerol-3-phosphate O-acyltransferase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000336984.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375104.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000375107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395497.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000395499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000476663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000476663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000476663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000476663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000476663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000476663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000490711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 ENST00000617181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_006411.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011481.hg.1 NM_032741.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0