This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00964411_m1
See other STX1A GE Assays ›
Gene Symbol:
STX1A
Gene Name:
syntaxin 1A
Gene Aliases:
HPC-1, P35-1, STX1, SYN1A
Chromosome Location:
Chr.7: 73699205 - 73719702 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00964411_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STX1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165903.1 NP_001159375.1 7 - 8 593 54
NM_004603.3 NP_004594.1 7 - 8 593 54
XM_017012567.1 XP_016868056.1 7 - 8 608 54
GenBank mRNA AB086954.1 - 7 - 8 537 54
AK299959.1 - 4 - 5 294 54
AK313980.1 - 7 - 8 541 54
BC003011.1 - 7 - 8 565 2820851 54
BC064644.1 - 7 - 8 564 5769528 54
CR457018.1 - 7 - 8 537 54
D37932.1 - 7 - 8 537 54
KJ534952.1 - 7 - 8 537 54
KJ534980.1 - 6 - 7 417 54
L37792.1 - 7 - 8 538 54
U12918.1 - 6 - 7 496 54

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STX1A

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of neurotransmitter secretion
intracellular protein transport
neurotransmitter secretion
response to gravity
positive regulation of norepinephrine secretion
regulation of synaptic vesicle priming
glutamate secretion
synaptic vesicle docking
synaptic vesicle fusion to presynaptic active zone membrane
secretion by cell
positive regulation of catecholamine secretion
positive regulation of calcium ion-dependent exocytosis
vesicle docking
regulation of insulin secretion
positive regulation of excitatory postsynaptic potential
SNARE binding
glycoprotein binding
SNAP receptor activity
protein binding
myosin binding
calcium channel inhibitor activity
chloride channel inhibitor activity
kinase binding
protein domain specific binding
protein binding, bridging
myosin head/neck binding
ATP-dependent protein binding
ion channel binding
protein heterodimerization activity
protein N-terminus binding
calcium-dependent protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0