Assay Details
Gene Symbol: CD47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001777.3 | NP_001768.1 | 7 - 8 | 1057 | 191 | |
NM_198793.2 | NP_942088.1 | 7 - 8 | 1057 | 133 | ||
XM_005247908.1 | XP_005247965.1 | 7 - 8 | 1027 | 158 | ||
GenBank mRNA | AI701980.1 | - | 3 - 4 | 158 | ||
AK289813.1 | - | 7 - 8 | 998 | 191 | ||
BC010016.2 | - | 7 - 8 | 959 | 4303534 | 133 | |
BC012884.1 | - | 7 - 8 | 980 | 3857911 | 133 | |
BC037306.1 | - | 7 - 8 | 994 | 5260986 | 133 | |
X69398.1 | - | 7 - 8 | 983 | 191 | ||
Z25521.1 | - | 7 - 8 | 1016 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001629.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
integrin-mediated signaling pathway
opsonization
positive regulation of cell proliferation
response to bacterium
positive regulation of cell-cell adhesion
extracellular matrix organization
positive regulation of inflammatory response
positive regulation of phagocytosis
positive regulation of T cell activation
leukocyte migration
protein binding
thrombospondin receptor activity
cell adhesion
integrin-mediated signaling pathway
opsonization
positive regulation of cell proliferation
response to bacterium
positive regulation of cell-cell adhesion
extracellular matrix organization
positive regulation of inflammatory response
positive regulation of phagocytosis
positive regulation of T cell activation
leukocyte migration
protein binding
thrombospondin receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03029387.hg.1 | ENST00000355354.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03029387.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03029387.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03029387.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03029387.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03029387.hg.1 | TC03001629.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300188799.hg.1 | TC0300011936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000355354.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000361309.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000398258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000471694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | ENST00000517766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_001777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011936.hg.1 | NM_198793.2 | Refseq |
