Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024032.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024032.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024032.hg.1 |
TC03003379.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013971.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013971.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013971.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013971.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013971.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013971.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024030.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024030.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024030.hg.1 |
TC03003379.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0300013971.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013971.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013971.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013971.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013971.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013971.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207343.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207343.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207343.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207343.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207343.hg.1 |
TC0300013971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300207341.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207341.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207341.hg.1 |
ENST00000273610.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207341.hg.1 |
NM_033199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207341.hg.1 |
TC0300013971.hg.1 |
Clariom D TC |