This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00963421_m1
See other PAPOLG GE Assays ›
Gene Symbol:
PAPOLG
Gene Name:
poly(A) polymerase gamma
Gene Aliases:
-
Chromosome Location:
Chr.2: 60756230 - 60802086 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00963421_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAPOLG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022894.3 NP_075045.2 15 - 16 1643 128
XM_005264500.3 XP_005264557.1 15 - 16 1643 128
XM_005264501.1 XP_005264558.1 15 - 16 1461 128
XM_011533041.1 XP_011531343.1 14 - 15 1349 128
XM_017004723.1 XP_016860212.1 14 - 15 1505 128
XM_017004724.1 XP_016860213.1 14 - 15 1505 128
GenBank mRNA AB209304.1 - 5 - 6 398 128
AF312211.1 - 15 - 16 1625 128
AJ308101.1 - 15 - 16 1394 128
AK021867.1 - 15 - 16 1579 128
AK023034.1 - 4 - 5 345 128
AK023544.1 - 14 - 15 1392 128
AK314663.1 - 15 - 16 1607 128
AY029162.1 - 15 - 16 1403 128
BC110906.1 - 15 - 16 1582 128
BC111701.1 - 15 - 16 1517 40038554 128
BC113747.1 - 15 - 16 1517 40038563 128
BX647824.1 - 14 - 15 1363 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200007702.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000347.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PAPOLG

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
RNA polyadenylation
RNA binding
polynucleotide adenylyltransferase activity
ATP binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02005150.hg.1 ENST00000238714.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02005150.hg.1 ENST00000378104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02005150.hg.1 NM_022894.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02005150.hg.1 TC02000347.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000238714.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000412217.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000414060.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000453839.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000470208.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000496283.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 NM_022894.3 Refseq
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000238714.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000412217.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000414060.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000453839.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000470208.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 ENST00000496283.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 NM_022894.3 Refseq
Affymetrix Human Clariom D Assay PSR0200153143.hg.1 TC0200007702.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0