This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00956740_m1
See other C11ORF63 GE Assays ›
Gene Symbol:
C11orf63
Gene Name:
chromosome 11 open reading frame 63
Gene Aliases:
-
Chromosome Location:
Chr.11: 122881640 - 122963862 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00956740_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C11orf63
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024806.3 NP_079082.2 3 - 4 1399 112
XM_005271680.4 XP_005271737.1 3 - 4 1260 112
XM_011543009.2 XP_011541311.1 3 - 4 1468 112
XM_017018338.1 XP_016873827.1 3 - 4 1134 112
XM_017018339.1 XP_016873828.1 3 - 4 1167 112
XM_017018340.1 XP_016873829.1 3 - 4 978 112
XM_017018341.1 XP_016873830.1 3 - 4 1749 112
XM_017018342.1 XP_016873831.1 3 - 4 1134 112
GenBank mRNA AK027207.1 - 2 - 3 753 112
AK291625.1 - 3 - 4 1002 112
BC068507.1 - 3 - 4 1162 5298472 112
CD244380.1 - 1 - 2 85 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100009335.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001117.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene C11orf63

Gene Ontology Categories:

Function(s)

brain development
ciliary basal body organization
cerebrospinal fluid secretion
axoneme assembly

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11014238.hg.1 ENST00000227349.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014238.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014238.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11014238.hg.1 TC11001117.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11014239.hg.1 ENST00000227349.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014239.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014239.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11014239.hg.1 TC11001117.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000534362.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000534362.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 ENST00000534362.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay PSR1100170082.hg.1 TC1100009335.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000227349.6 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000307257.10 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000531316.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000534362.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000534362.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 ENST00000534362.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_024806.3 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq
Affymetrix Human Clariom D Assay TC1100009335.hg.1 NM_199124.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm