This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00956646_m1
See other PODN GE Assays ›
Gene Symbol:
PODN
Gene Name:
podocan
Gene Aliases:
PCAN, SLRR5A
Chromosome Location:
Chr.1: 53062052 - 53085502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00956646_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PODN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153703.4 NP_714914.2 4 - 5 782 78
XM_005270457.3 XP_005270514.1 5 - 6 811 78
XM_017000294.1 XP_016855783.1 4 - 5 1132 78
GenBank mRNA AK296649.1 - 5 - 6 677 78
AK299375.1 - 6 - 7 867 78
AK314096.1 - 4 - 5 376 78
AY313607.1 - 4 - 5 621 78
AY358294.1 - 6 - 7 898 78
BC028292.1 - 4 - 5 605 4938380 78
BC030608.2 - 4 - 5 623 4826932 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008341.hg
This assay targets the same location as HTA transcript cluster(s) TC01000649.hg

Additional Information:

This assay is in the 5'UTR region of the AK314096.1,BC028292.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PODN

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
negative regulation of cell proliferation
cytokine-mediated signaling pathway
negative regulation of cell migration
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
collagen binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0