This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00954810_m1
See other CENPQ GE Assays ›
Gene Symbol:
CENPQ
Gene Name:
centromere protein Q
Gene Aliases:
C6orf139, CENP-Q
Chromosome Location:
Chr.6: 49463327 - 49493107 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00954810_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CENPQ
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018132.3 NP_060602.2 6 - 7 563 85
XM_005249205.1 XP_005249262.1 6 - 7 624 85
XM_011514723.2 XP_011513025.2 5 - 6 499 85
GenBank mRNA AK001407.1 - 1 - 2 113 85
AK293016.1 - 6 - 7 568 85
BC016279.1 - 6 - 7 553 3908895 85
BI768787.1 - 6 - 7 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008209.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000640.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CENPQ

Gene Ontology Categories:

Function(s)

sister chromatid cohesion
CENP-A containing nucleosome assembly

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007784.hg.1 ENST00000335783.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007784.hg.1 ENST00000371200.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007784.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007784.hg.1 TC06000640.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay PSR0600157423.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay PSR0600157423.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay PSR0600157423.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay PSR0600157423.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay PSR0600157423.hg.1 TC0600008209.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0