This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00954809_m1
See other CENPQ GE Assays ›
Gene Symbol:
CENPQ
Gene Name:
centromere protein Q
Gene Aliases:
C6orf139, CENP-Q
Chromosome Location:
Chr.6: 49463327 - 49493107 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00954809_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CENPQ
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018132.3 NP_060602.2 5 - 6 438 149
XM_005249205.1 XP_005249262.1 5 - 6 499 149
XM_011514723.2 XP_011513025.2 4 - 5 374 149
GenBank mRNA AK293016.1 - 5 - 6 443 149
BC016279.1 - 5 - 6 428 3908895 149
BI768787.1 - 5 - 6 457 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008209.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000640.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CENPQ

Gene Ontology Categories:

Function(s)

sister chromatid cohesion
CENP-A containing nucleosome assembly

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06007783.hg.1 ENST00000335783.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007783.hg.1 ENST00000371200.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007783.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007783.hg.1 TC06000640.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay TC0600008209.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay PSR0600157422.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay PSR0600157422.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay PSR0600157422.hg.1 ENST00000335783.3 Ensembl
Affymetrix Human Clariom D Assay PSR0600157422.hg.1 NM_018132.3 Refseq
Affymetrix Human Clariom D Assay PSR0600157422.hg.1 TC0600008209.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0