Assay Details
Gene Symbol: NECAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019065.2 | NP_061938.2 | 2 - 3 | 233 | 84 | |
XM_011523135.1 | XP_011521437.1 | 2 - 3 | 76 | 84 | ||
GenBank mRNA | AK127376.1 | - | 2 - 3 | 154 | 84 | |
AY299331.1 | - | 2 - 3 | 233 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000653.hg
Additional Information:
This assay is in the 5'UTR region of the XM_011523135.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16009193.hg.1 | ENST00000305202.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16009193.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16009193.hg.1 | TC16000653.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16009194.hg.1 | ENST00000305202.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16009194.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16009194.hg.1 | TC16000653.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000564166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | ENST00000567703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008633.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | ENST00000565691.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600164444.hg.1 | TC1600008633.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | ENST00000305202.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | ENST00000566836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | NM_019065.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600164446.hg.1 | TC1600008633.hg.1 | Clariom D TC |
