This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00950769_m1
See other AGR2 GE Assays ›
Gene Symbol:
AGR2
Gene Name:
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases:
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
Chromosome Location:
Chr.7: 16791811 - 16805114 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00950769_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006408.3 NP_006399.1 5 - 6 509 70
XM_005249581.4 XP_005249638.1 5 - 6 478 70
GenBank mRNA AF007791.1 - 4 - 5 332 70
AF038451.1 - 5 - 6 380 70
AF087879.1 - 5 - 6 471 70
AF088867.1 - 5 - 6 469 70
AF115926.1 - 5 - 6 379 70
AK223067.1 - 5 - 6 400 70
AK312942.1 - 5 - 6 390 70
AY359009.1 - 5 - 6 469 70
BC015503.1 - 5 - 6 378 3852448 70
BT007048.1 - 4 - 5 322 70
FJ224307.1 - 4 - 5 332 70
KJ767789.1 - 5 - 6 483 70
KJ767790.1 - 2 - 3 101 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010355.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001163.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AGR2

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of gene expression
positive regulation of cell-substrate adhesion
response to endoplasmic reticulum stress
positive regulation of epidermal growth factor receptor signaling pathway
digestive tract morphogenesis
positive regulation of developmental growth
lung goblet cell differentiation
negative regulation of cell death
mucus secretion
positive regulation of establishment of protein localization to plasma membrane
positive regulation of IRE1-mediated unfolded protein response
positive regulation of PERK-mediated unfolded protein response
dystroglycan binding
epidermal growth factor receptor binding
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07018196.hg.1 ENST00000401412.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018196.hg.1 ENST00000419304.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018196.hg.1 ENST00000419572.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018196.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07018196.hg.1 TC07001163.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay PSR0700173609.hg.1 TC0700010355.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0