This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00950507_m1
See other GIF GE Assays ›
Gene Symbol:
GIF
Gene Name:
gastric intrinsic factor
Gene Aliases:
IF, IFMH, INF, TCN3
Chromosome Location:
Chr.11: 59829268 - 59845529 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00950507_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GIF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005142.2 NP_005133.2 6 - 7 920 82
GenBank mRNA AK301295.1 - 6 - 7 1032 82
AK314275.1 - 6 - 7 939 82
BC037958.1 - 6 - 7 920 5759934 82
X76562.1 - 6 - 7 919 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100010991.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001836.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GIF

Gene Ontology Categories:

Function(s) Process(es)

cobalt ion transport
cobalamin metabolic process
cobalamin transport
cobalamin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header