This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00947875_m1
See other TGFBR2 GE Assays ›
Gene Symbol:
TGFBR2
Gene Name:
transforming growth factor beta receptor 2
Gene Aliases:
AAT3, FAA3, LDS1B, LDS2, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII
Chromosome Location:
Chr.3: 30606490 - 30694142 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00947875_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGFBR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024847.2 NP_001020018.1 1 - 2 475 93
GenBank mRNA AK300383.1 - 1 - 2 376 93
CB988859.1 - 1 - 2 403 93
D28131.1 - 1 - 2 33 93
KU178360.1 - 1 - 2 93 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300006925.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000149.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TGFBR2

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

blood vessel development
patterning of blood vessels
vasculogenesis
in utero embryonic development
heart looping
positive regulation of mesenchymal cell proliferation
lens development in camera-type eye
positive regulation of tolerance induction to self antigen
positive regulation of B cell tolerance induction
positive regulation of T cell tolerance induction
outflow tract morphogenesis
growth plate cartilage chondrocyte growth
protein phosphorylation
receptor-mediated endocytosis
apoptotic process
transforming growth factor beta receptor signaling pathway
common-partner SMAD protein phosphorylation
Notch signaling pathway
smoothened signaling pathway
gastrulation
brain development
heart development
embryo implantation
aging
response to nutrient
positive regulation of cell proliferation
response to mechanical stimulus
response to glucose
regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of epithelial to mesenchymal transition
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
negative regulation of transforming growth factor beta receptor signaling pathway
organ regeneration
activation of protein kinase activity
embryonic hemopoiesis
wound healing
regulation of cell proliferation
response to drug
myeloid dendritic cell differentiation
positive regulation of skeletal muscle tissue regeneration
response to estrogen
positive regulation of angiogenesis
response to steroid hormone
digestive tract development
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
positive regulation of NK T cell differentiation
palate development
negative regulation of cardiac muscle cell proliferation
pathway-restricted SMAD protein phosphorylation
ventricular septum morphogenesis
bronchus morphogenesis
trachea formation
mammary gland morphogenesis
lung lobe morphogenesis
response to cholesterol
positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
lens fiber cell apoptotic process
positive regulation of reactive oxygen species metabolic process
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta-activated receptor activity
transforming growth factor beta receptor activity, type II
protein binding
ATP binding
glycosaminoglycan binding
mitogen-activated protein kinase kinase kinase binding
type I transforming growth factor beta receptor binding
SMAD binding
metal ion binding
transforming growth factor beta binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0