Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000591614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
ENST00000591614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022190.hg.1 |
ENST00000323040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022190.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022190.hg.1 |
TC19002713.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022192.hg.1 |
ENST00000323040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022192.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022192.hg.1 |
TC19002713.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022191.hg.1 |
ENST00000323040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022191.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022191.hg.1 |
TC19002713.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900206805.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206805.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206805.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206805.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206805.hg.1 |
TC1900011983.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
ENST00000591614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
ENST00000591614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206806.hg.1 |
TC1900011983.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900206804.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206804.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206804.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206804.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206804.hg.1 |
TC1900011983.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900206808.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206808.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206808.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206808.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206808.hg.1 |
TC1900011983.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000323040.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000591614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
ENST00000591614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011983.hg.1 |
NM_005282.2 |
Refseq |