This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00947723_m1
See other CELSR1 GE Assays ›
Gene Symbol:
CELSR1
Gene Name:
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases:
ADGRC1, CDHF9, FMI2, HFMI2, ME2
Chromosome Location:
Chr.22: 46360834 - 46537502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00947723_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CELSR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014246.1 NP_055061.1 9 - 10 5226 64
XM_006724383.3 XP_006724446.1 9 - 10 5553 64
XM_011530553.1 XP_011528855.1 9 - 10 5558 64
XM_011530554.2 XP_011528856.1 9 - 10 1769 64
XM_011530555.2 XP_011528857.1 9 - 10 1850 64
GenBank mRNA AF231024.1 - 9 - 10 5226 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200009021.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000889.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CELSR1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

establishment of planar polarity
neuron migration
neural tube closure
hair follicle development
homophilic cell adhesion via plasma membrane adhesion molecules
G-protein coupled receptor signaling pathway
Rho protein signal transduction
central nervous system development
locomotory behavior
anterior/posterior pattern specification
regulation of actin cytoskeleton organization
wound healing
establishment of planar polarity of embryonic epithelium
inner ear morphogenesis
apical protein localization
establishment of body hair planar orientation
Wnt signaling pathway, planar cell polarity pathway
orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis
planar dichotomous subdivision of terminal units involved in lung branching morphogenesis
lateral sprouting involved in lung morphogenesis
planar cell polarity pathway involved in neural tube closure
protein localization involved in establishment of planar polarity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000468025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000468025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000468025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000497509.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 ENST00000497509.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22014948.hg.1 ENST00000262738.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22014948.hg.1 NM_014246.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22014948.hg.1 TC22000889.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000454637.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000468025.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000468025.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000468025.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000473624.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000497509.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 ENST00000497509.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay TC2200009021.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay PSR2200165369.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200165369.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay PSR2200165369.hg.1 ENST00000262738.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200165369.hg.1 NM_014246.1 Refseq
Affymetrix Human Clariom D Assay PSR2200165369.hg.1 TC2200009021.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0