This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00946874_g1
See other GDPD3 GE Assays ›
Gene Symbol:
GDPD3
Gene Name:
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases:
GDE7
Chromosome Location:
Chr.16: 30104810 - 30113557 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00946874_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDPD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK026256.1 - 1 - 2 199 129
AK225450.1 - 1 - 2 199 129

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK026256.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566613.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566613.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 ENST00000566613.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000360688.3 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000406256.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000565704.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566434.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566613.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566613.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 ENST00000566613.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq
Affymetrix Human Clariom D Assay TC1600009953.hg.1 NM_024307.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0