Assay Details
Gene Symbol: GDPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024307.2 | NP_077283.2 | 8 - 9 | 843 | 68 | |
GenBank mRNA | AK026256.1 | - | 7 - 8 | 1051 | 68 | |
AK225450.1 | - | 7 - 8 | 1051 | 68 | ||
BC002714.2 | - | 8 - 9 | 843 | 3629566 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001020.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16014584.hg.1 | ENST00000360688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16014584.hg.1 | ENST00000406256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16014584.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16014584.hg.1 | TC16001020.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600176754.hg.1 | TC1600009953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
