Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016202.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016202.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016202.hg.1 |
TC11001277.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016201.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016201.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016201.hg.1 |
TC11001277.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016200.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016200.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016200.hg.1 |
TC11001277.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009791.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174009.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174009.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174009.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174009.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174009.hg.1 |
TC1100009791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174010.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174010.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174010.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174010.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174010.hg.1 |
TC1100009791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174011.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174011.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174011.hg.1 |
ENST00000331289.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174011.hg.1 |
NM_005170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174011.hg.1 |
TC1100009791.hg.1 |
Clariom D TC |