Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022049.hg.1 |
ENST00000252482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022049.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022049.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022049.hg.1 |
TC19001616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022050.hg.1 |
ENST00000252482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022050.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022050.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022050.hg.1 |
TC19001616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190676.hg.1 |
TC1900010931.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190677.hg.1 |
TC1900010931.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000252482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
ENST00000413988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010931.hg.1 |
NM_138568.4 |
Refseq |