This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00943233_g1
See other LTBP4 GE Assays ›
Gene Symbol:
LTBP4
Gene Name:
latent transforming growth factor beta binding protein 4
Gene Aliases:
ARCL1C, LTBP-4, LTBP4L, LTBP4S
Chromosome Location:
Chr.19: 40592888 - 40629820 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00943233_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LTBP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003573.2 NP_003564.2 2 - 3 75 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LTBP4

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
protein folding
transmembrane receptor protein serine/threonine kinase signaling pathway
multicellular organism development
regulation of transforming growth factor beta receptor signaling pathway
regulation of proteolysis
growth hormone secretion
regulation of cell differentiation
transforming growth factor beta-activated receptor activity
integrin binding
calcium ion binding
protein binding
glycosaminoglycan binding
transforming growth factor beta binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0