This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00942485_g1
See other IFITM5 GE Assays ›
Gene Symbol:
IFITM5
Gene Name:
interferon induced transmembrane protein 5
Gene Aliases:
BRIL, DSPA1, Hrmp1, OI5, fragilis4
Chromosome Location:
Chr.11: 298200 - 299526 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00942485_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFITM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025295.2 NP_001020466.1 1 - 2 227 58
GenBank mRNA BC150562.1 - 1 - 2 220 40127610 58
BC150563.1 - 1 - 2 220 40127611 58
CR747200.1 - 1 - 2 229 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013119.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003502.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IFITM5

Gene Ontology Categories:

Function(s)

in utero embryonic development
response to biotic stimulus
bone mineralization
regulation of bone mineralization
bone morphogenesis

MicroArray Probes:

Agilent Affymetrix ArrayStar Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594354 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594335 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594347 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594342 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988164 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594339 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594351 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594338 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988152 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988145 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594346 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594345 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594353 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594349 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594344 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594336 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594352 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594355 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988151 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07594343 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988147 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988155 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27988148 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3352995 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P150664 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 43673 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28585 - -
Agilent Human Genome CGH + SNP Microarray A_16_P02378583 - -
Agilent Human Genome CGH + SNP Microarray A_16_P02378581 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3352995 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3352995 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594349 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594351 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594354 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988153 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594343 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594338 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988172 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594350 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988164 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594347 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594352 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594355 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988155 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594345 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594342 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988151 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594347 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594349 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594345 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594355 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594354 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988153 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988155 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594352 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594343 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594338 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988151 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594350 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988164 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594351 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988172 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594342 - -
Whole Human Genome Oligo Microarray G4112A A_23_P150664 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594356 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594357 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594363 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594354 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594358 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594364 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P07594360 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405590 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405591 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594349 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405589 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405582 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594357 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594351 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594350 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405581 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P150664 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02378583 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594338 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405574 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594336 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405590 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594344 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405522 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594304 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594303 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594315 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594316 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594341 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594355 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405533 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594357 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594320 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594313 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594347 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405545 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405583 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594314 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594349 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405570 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594307 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405519 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594311 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594301 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405531 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594364 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594306 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594360 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594340 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594342 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594363 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405517 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405539 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405541 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594354 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405579 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594346 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405546 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405584 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405525 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405582 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594343 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594308 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07594358 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16405532 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_001025295_at - -
Affymetrix Human Gene 1.0 ST Array 7945370 - -
Affymetrix Human Gene 1.0 ST Array 7945369 - -
Affymetrix Human Gene 1.1 ST Array 7945368 - -
Affymetrix Human Genome U219 Array 11737383_at - -
Affymetrix Human Genome U219 Array 11737383_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR11015373.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015373.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015373.hg.1 TC11003502.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203125.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100203125.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203125.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100203125.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203125.hg.1 TC1100013119.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A044710 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A044710 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1711032 NM_001025295.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1711032 BC150562.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1720992 - -
HumanHT-12 V4.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_27022-S - -
Illumina human-6 v2.0 expression beadchip ILMN_1720992 - -
Illumina humanRef-8 v2.0 expression beadchip ILMN_1720992 - -
Illumina Sentrix Human-6 v2 Expression BeadChip 5560162 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_27022 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1720992 - -
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711032 NM_001025295.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711032 BC150562.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711032 BC150563.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0