This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00941707_m1
See other PDGFD GE Assays ›
Gene Symbol:
PDGFD
Gene Name:
platelet derived growth factor D
Gene Aliases:
IEGF, MSTP036, SCDGF-B, SCDGFB
Chromosome Location:
Chr.11: 103907186 - 104164299 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00941707_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDGFD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025208.4 NP_079484.1 1 - 2 497 63
GenBank mRNA AB033832.1 - 1 - 2 281 63
AF113216.1 - 1 - 2 231 63
AF335584.1 - 1 - 2 306 63
AF336376.1 - 1 - 2 300 63
AK292801.1 - 1 - 2 305 63
AY027517.1 - 1 - 2 217 63
BG722948.1 - 1 - 2 368 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDGFD

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
blood coagulation
positive regulation of cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of cell migration
positive regulation of protein autophosphorylation
cellular response to platelet-derived growth factor stimulus
positive regulation of MAP kinase activity
platelet-derived growth factor receptor signaling pathway
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell division
cellular response to hydrogen peroxide
positive regulation of ERK1 and ERK2 cascade
cellular response to amino acid stimulus
cellular response to transforming growth factor beta stimulus
positive regulation of smooth muscle cell chemotaxis
positive regulation of glomerular mesangial cell proliferation
positive regulation of monocyte extravasation
platelet-derived growth factor receptor binding
growth factor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000302251.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000393158.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 ENST00000529268.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_025208.4 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Affymetrix Human Clariom D Assay TC1100012155.hg.1 NM_033135.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 108301 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0