This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00941444_m1
See other PPP4R4 GE Assays ›
Gene Symbol:
PPP4R4
Gene Name:
protein phosphatase 4 regulatory subunit 4
Gene Aliases:
CFAP14, KIAA1622, PP4R4
Chromosome Location:
Chr.14: 94174312 - 94279735 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00941444_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP4R4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_058237.1 NP_478144.1 22 - 23 2529 153
XM_005267930.2 XP_005267987.1 22 - 23 2362 153
XM_011537038.2 XP_011535340.1 22 - 23 2525 153
XM_011537039.2 XP_011535341.1 21 - 22 2270 153
XM_011537040.2 XP_011535342.1 20 - 21 2177 153
XM_017021528.1 XP_016877017.1 20 - 21 2214 153
XM_017021529.1 XP_016877018.1 24 - 25 2884 153
XM_017021530.1 XP_016877019.1 24 - 25 2858 153
GenBank mRNA AB046842.1 - 22 - 23 2448 153
AK303722.1 - 10 - 11 1011 153
BC068491.1 - 22 - 23 2488 5297335 153

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008058.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000585.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP4R4

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphoprotein phosphatase activity
regulation of protein serine/threonine phosphatase activity
protein binding
protein phosphatase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14007242.hg.1 ENST00000304338.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14007242.hg.1 NM_058237.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14007242.hg.1 TC14000585.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000328839.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000553661.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000555690.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556470.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 ENST00000556884.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_020958.2 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay TC1400008058.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay PSR1400157193.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400157193.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay PSR1400157193.hg.1 ENST00000304338.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400157193.hg.1 NM_058237.1 Refseq
Affymetrix Human Clariom D Assay PSR1400157193.hg.1 TC1400008058.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0