Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000346743.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000400304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000400308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000400309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000400310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000400314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000449640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
ENST00000468429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
NM_001130141.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
NM_020528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002985.hg.1 |
TC21000247.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400310.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000449640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000468429.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000475402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000626237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
NM_001130141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
NM_020528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000400310.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000449640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000468429.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000475402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
ENST00000626237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
NM_001130141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
NM_020528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158503.hg.1 |
TC2100008536.hg.1 |
Clariom D TC |