Assay Details
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC110337.1 | - | 5 - 6 | 581 | 5755200 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179185.hg.1 | TC0600011553.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179187.hg.1 | TC0600011553.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011553.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | ENST00000360661.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | ENST00000374467.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | ENST00000442998.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | NM_001188.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600179186.hg.1 | TC0600011553.hg.1 | Clariom D TC |
