Assay Details
Gene Symbol: ADGRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032777.9 | NP_116166.9 | 17 - 18 | 3043 | 97 | |
GenBank mRNA | AB040964.2 | - | 14 - 15 | 2369 | 97 | |
AF378755.1 | - | 17 - 18 | 2670 | 97 | ||
AK074099.2 | - | 17 - 18 | 2574 | 97 | ||
AL832524.1 | - | 1 - 2 | 110 | 97 | ||
AL832959.1 | - | 10 - 11 | 1713 | 97 | ||
AY181242.1 | - | 17 - 18 | 2636 | 97 | ||
BC146774.1 | - | 14 - 15 | 2369 | 9007223 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000279.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
central nervous system development
positive regulation of endothelial cell migration
endothelial cell migration
regulation of angiogenesis
regulation of chemotaxis
regulation of establishment of blood-brain barrier
negative regulation of vascular endothelial growth factor signaling pathway
G-protein coupled receptor activity
protein binding
sprouting angiogenesis
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
central nervous system development
positive regulation of endothelial cell migration
endothelial cell migration
regulation of angiogenesis
regulation of chemotaxis
regulation of establishment of blood-brain barrier
negative regulation of vascular endothelial growth factor signaling pathway
G-protein coupled receptor activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08003898.hg.1 | ENST00000315215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003898.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003898.hg.1 | ENST00000416514.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003898.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003898.hg.1 | TC08000279.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150240.hg.1 | TC0800007311.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
