Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000523248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000523248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001230.hg.1 |
ENST00000251630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001230.hg.1 |
ENST00000398074.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001230.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001230.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001230.hg.1 |
TC08000128.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000523248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000523248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006873.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
ENST00000251630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
ENST00000541323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
NM_006207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146531.hg.1 |
TC0800006873.hg.1 |
Clariom D TC |