Assay Details
Gene Symbol: TMEM9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073431.1 | 3 - 4 | 733 | 68 | ||
GenBank mRNA | DA631400.1 | - | 3 - 4 | 510 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147689.hg.1 | TC1100006798.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006798.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147691.hg.1 | TC1100006798.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147692.hg.1 | TC1100006798.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000525484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000532599.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | ENST00000534349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | NR_073431.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100147687.hg.1 | TC1100006798.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 210759 | - | - |
