This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00936627_g1
See other ADGRA2 GE Assays ›
Gene Symbol:
ADGRA2
Gene Name:
adhesion G protein-coupled receptor A2
Gene Aliases:
GPR124, TEM5
Chromosome Location:
Chr.8: 37796883 - 37843986 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00936627_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032777.9 NP_116166.9 9 - 10 1680 58
XM_005273471.3 XP_005273528.1 9 - 10 1680 58
XM_011544481.2 XP_011542783.1 9 - 10 1680 58
XM_011544482.2 XP_011542784.1 8 - 9 1608 58
XM_011544483.2 XP_011542785.1 9 - 10 1680 58
GenBank mRNA AB040964.2 - 9 - 10 1657 58
AF378755.1 - 9 - 10 1307 58
AK074099.2 - 9 - 10 1211 58
AL832959.1 - 2 - 3 350 58
AY181242.1 - 9 - 10 1273 58
BC146774.1 - 9 - 10 1657 9007223 58
KU257672.1 - 9 - 10 1192 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800007311.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000279.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRA2

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

sprouting angiogenesis
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
central nervous system development
positive regulation of endothelial cell migration
endothelial cell migration
regulation of angiogenesis
regulation of chemotaxis
regulation of establishment of blood-brain barrier
negative regulation of vascular endothelial growth factor signaling pathway
G-protein coupled receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08003887.hg.1 ENST00000315215.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08003887.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08003887.hg.1 ENST00000416514.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08003887.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08003887.hg.1 TC08000279.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 ENST00000428068.5 Ensembl
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay TC0800007311.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 ENST00000315215.11 Ensembl
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 ENST00000412232.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 NM_032777.9 Refseq
Affymetrix Human Clariom D Assay PSR0800150232.hg.1 TC0800007311.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0