Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003884.hg.1 |
ENST00000315215.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003884.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003884.hg.1 |
ENST00000416514.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003884.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003884.hg.1 |
TC08000279.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
ENST00000428068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007311.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
ENST00000315215.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
ENST00000412232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
NM_032777.9 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150229.hg.1 |
TC0800007311.hg.1 |
Clariom D TC |