Assay Details
Gene Symbol: ADGRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032777.9 | NP_116166.9 | 13 - 14 | 2435 | 89 | |
XM_005273471.3 | XP_005273528.1 | 13 - 14 | 2435 | 89 | ||
XM_011544481.2 | XP_011542783.1 | 13 - 14 | 2435 | 89 | ||
XM_011544482.2 | XP_011542784.1 | 12 - 13 | 2363 | 89 | ||
XM_011544483.2 | XP_011542785.1 | 13 - 14 | 2435 | 89 | ||
GenBank mRNA | AF378755.1 | - | 13 - 14 | 2062 | 89 | |
AK074099.2 | - | 13 - 14 | 1966 | 89 | ||
AL832959.1 | - | 6 - 7 | 1105 | 89 | ||
AY181242.1 | - | 13 - 14 | 2028 | 89 | ||
KU257672.1 | - | 13 - 14 | 1947 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000279.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
central nervous system development
positive regulation of endothelial cell migration
endothelial cell migration
regulation of angiogenesis
regulation of chemotaxis
regulation of establishment of blood-brain barrier
negative regulation of vascular endothelial growth factor signaling pathway
G-protein coupled receptor activity
protein binding
sprouting angiogenesis
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
central nervous system development
positive regulation of endothelial cell migration
endothelial cell migration
regulation of angiogenesis
regulation of chemotaxis
regulation of establishment of blood-brain barrier
negative regulation of vascular endothelial growth factor signaling pathway
G-protein coupled receptor activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003891.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003891.hg.1 | ENST00000416514.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003891.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003891.hg.1 | TC08000279.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003893.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003893.hg.1 | ENST00000416514.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003893.hg.1 | ENST00000521191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003893.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003893.hg.1 | TC08000279.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000315215.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | ENST00000428068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007311.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150236.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150236.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150236.hg.1 | ENST00000412232.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150236.hg.1 | NM_032777.9 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150236.hg.1 | TC0800007311.hg.1 | Clariom D TC |
