This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00936139_m1
See other TULP4 GE Assays ›
Gene Symbol:
TULP4
Gene Name:
tubby like protein 4
Gene Aliases:
TUSP
Chromosome Location:
Chr.6: 158232192 - 158511828 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00936139_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TULP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007466.2 NP_001007467.1 2 - 3 1738 61
NM_020245.4 NP_064630.2 2 - 3 1738 61
XM_011535946.1 XP_011534248.1 3 - 4 2415 61
XM_017011063.1 XP_016866552.1 4 - 5 2564 61
XM_017011064.1 XP_016866553.1 5 - 6 2729 61
XM_017011065.1 XP_016866554.1 4 - 5 2482 61
XM_017011066.1 XP_016866555.1 3 - 4 2372 61
XM_017011067.1 XP_016866556.1 3 - 4 2465 61
XM_017011068.1 XP_016866557.1 3 - 4 2431 61
XM_017011069.1 XP_016866558.1 4 - 5 2567 61
XM_017011070.1 XP_016866559.1 3 - 4 2415 61
XM_017011071.1 XP_016866560.1 4 - 5 2567 61
GenBank mRNA AB037818.1 - 2 - 3 1738 61
AF219946.2 - 2 - 3 1738 61
AF288480.1 - 2 - 3 1738 61
AK304056.1 - 2 - 3 953 61
BC152476.1 - 2 - 3 1738 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600014198.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001132.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TULP4

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
protein ubiquitination
intracellular signal transduction
protein localization to cilium
transcription factor activity, sequence-specific DNA binding
phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000461853.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000613994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06013472.hg.1 ENST00000367094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06013472.hg.1 ENST00000367097.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06013472.hg.1 NM_001007466.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06013472.hg.1 NM_020245.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06013472.hg.1 TC06001132.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay PSR0600199822.hg.1 TC0600014198.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367094.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000367097.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000461853.3 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613390.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613994.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000613994.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 ENST00000616856.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_001007466.2 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq
Affymetrix Human Clariom D Assay TC0600014198.hg.1 NM_020245.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0