This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00935712_g1
See other TTLL8 GE Assays ›
Gene Symbol:
TTLL8
Gene Name:
tubulin tyrosine ligase like 8
Gene Aliases:
-
Chromosome Location:
Chr.22: 50018575 - 50058227 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00935712_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTLL8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017029176.1 XP_016884665.1 7 - 8 725 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein polyglycylation
ATP binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000477219.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0