Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
ENST00000531627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
ENST00000531627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179346.hg.1 |
ENST00000531627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179346.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179346.hg.1 |
ENST00000531627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179346.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179346.hg.1 |
TC1100010461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
ENST00000531627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
ENST00000531627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
ENST00000531962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010461.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179345.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179345.hg.1 |
NR_120527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179345.hg.1 |
TC1100010461.hg.1 |
Clariom D TC |