This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00932282_g1
See other SIM1 GE Assays ›
Gene Symbol:
SIM1
Gene Name:
single-minded family bHLH transcription factor 1
Gene Aliases:
bHLHe14
Chromosome Location:
Chr.6: 100385009 - 100464929 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00932282_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005068.2 NP_005059.2 6 - 7 953 99
XM_005267100.2 XP_005267157.1 7 - 8 1529 99
XM_011536072.2 XP_011534374.1 7 - 8 1529 99
XM_017011197.1 XP_016866686.1 7 - 8 1261 99
GenBank mRNA AK308090.1 - 7 - 8 1529 99
U70212.1 - 6 - 7 962 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012670.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001969.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SIM1

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

ureteric bud development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nervous system development
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000505753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000505753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06024371.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06024371.hg.1 ENST00000369208.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06024371.hg.1 NM_005068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06024371.hg.1 TC06001969.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000505753.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000505753.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 ENST00000511871.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay TC0600012670.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 ENST00000262901.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 ENST00000369208.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 NM_005068.2 Refseq
Affymetrix Human Clariom D Assay PSR0600187375.hg.1 TC0600012670.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0