Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017979.hg.1 |
ENST00000374647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017979.hg.1 |
ENST00000467959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017979.hg.1 |
ENST00000537196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017979.hg.1 |
NM_003640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017979.hg.1 |
TC09001455.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000374647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000467959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000495759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000537196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
NM_003640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000374647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000467959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000495759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
ENST00000537196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
NM_003640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175768.hg.1 |
TC0900011146.hg.1 |
Clariom D TC |