This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00929908_g1
See other RPL17 GE Assays ›
Gene Symbol:
RPL17
Gene Name:
ribosomal protein L17
Gene Aliases:
L17, PD-1, RPL23
Chromosome Location:
Chr.18: 49488481 - 49492565 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00929908_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000985.4 NP_000976.1 6 - 7 878 79
NM_001035006.2 NP_001030178.1 6 - 7 629 79
NM_001199340.1 NP_001186269.1 7 - 8 727 79
NM_001199341.1 NP_001186270.1 6 - 7 728 79
NM_001199342.1 NP_001186271.1 7 - 8 826 79
NM_001199343.1 NP_001186272.1 7 - 8 809 79
NM_001199344.1 NP_001186273.1 6 - 7 711 79
NM_001199345.1 NP_001186274.1 5 - 6 588 79
GenBank mRNA AK304659.1 - 5 - 6 502 79
BC000502.2 - 5 - 6 781 2821464 79
BC017831.1 - 1 547 4251433 79
BC066323.1 - 1 543 4797583 79
BC066324.1 - 1 547 4799029 79
BC106031.1 - 1 514 4540700 79
BQ882430.1 - 6 - 7 693 79
X53777.1 - 5 - 6 646 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL17

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0